Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(4-methoxyphenyl)pyridazin-3-amine | 4776-87-8 | MFCD06801353 | 97.3% | 201.23 | C11H11N3O | 1 MG
SDP

Catalog No. 5000301611
Encompass_Preferred

6-(4-methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic intermediate in medicinal chemistry research. It is supplied as a light brown solid of high purity for use in organic synthesis and assay development. Intended for research use only.

  • High purity: 97.3%.
  • Molecular formula: C11H11N3O.
  • Molecular weight: 201.23 g/mol.
  • Appearance: light brown to brown solid.
  • Intended for research use and synthetic applications.
  • Suitable as an intermediate for GABA-A receptor antagonist synthesis.

Catalog No. 50-003-01611 Supplier Medchemexpress LLC Supplier No. HYW0254431MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-(4-methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic intermediate in medicinal chemistry research. It is supplied as a light brown solid of high purity for use in organic synthesis and assay development. Intended for research use only.

  • High purity: 97.3%.
  • Molecular formula: C11H11N3O.
  • Molecular weight: 201.23 g/mol.
  • Appearance: light brown to brown solid.
  • Intended for research use and synthetic applications.
  • Suitable as an intermediate for GABA-A receptor antagonist synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.