Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(4-methoxyphenyl)pyridazin-3-amine | 4776-87-8 | MFCD06801353 | 97.3% | 201.23 | C11H11N3O | 10 MG
SDP

Catalog No. 5000299641
Encompass_Preferred

6-(4-methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic building block in medicinal chemistry, notably for the synthesis of SR 95103, a selective competitive GABA-A receptor antagonist. The compound is provided as a solid for research use with documented purity, formula, and solubility suitable for in vitro work.

  • High stated purity: 97.3%.
  • Chemical formula C11H11N3O.
  • Molecular weight 201.23 g/mol.
  • Supplied as a light brown to brown solid.
  • Soluble in DMSO at ≥ 15 mg/mL.
  • Recommended storage: powder -20°C for long-term, 4°C for shorter term.
  • Available in small research pack sizes, including 10 mg.

Catalog No. 50-002-99641 Supplier Medchemexpress LLC Supplier No. HYW02544310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

6-(4-methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic building block in medicinal chemistry, notably for the synthesis of SR 95103, a selective competitive GABA-A receptor antagonist. The compound is provided as a solid for research use with documented purity, formula, and solubility suitable for in vitro work.

  • High stated purity: 97.3%.
  • Chemical formula C11H11N3O.
  • Molecular weight 201.23 g/mol.
  • Supplied as a light brown to brown solid.
  • Soluble in DMSO at ≥ 15 mg/mL.
  • Recommended storage: powder -20°C for long-term, 4°C for shorter term.
  • Available in small research pack sizes, including 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.