Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(4-methoxyphenyl)pyridazin-3-amine | 4776-87-8 | MFCD06801353 | 97.3% | 201.23 g·mol^-1 | C11H11N3O | 25 MG
SDP

Catalog No. 5000302975
Encompass_Preferred

6-(4-Methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic intermediate in medicinal chemistry and research on GABA-A receptor antagonists. It is provided as a light brown solid with high stated purity and is intended for research and synthesis applications.

  • High purity (97.3%) suitable for analytical and synthetic work.
  • Molecular formula C11H11N3O; molecular weight 201.23 g·mol^-1.
  • Available in multiple small pack sizes for screening and synthesis.
  • Supplied as a solid, light brown to brown, stable under recommended storage.
  • Analytical documentation available: data sheet, COA, HNMR, LCMS, and SDS.

Catalog No. 50-003-02975 Supplier Medchemexpress LLC Supplier No. 25MGHYW025443
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-(4-Methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic intermediate in medicinal chemistry and research on GABA-A receptor antagonists. It is provided as a light brown solid with high stated purity and is intended for research and synthesis applications.

  • High purity (97.3%) suitable for analytical and synthetic work.
  • Molecular formula C11H11N3O; molecular weight 201.23 g·mol^-1.
  • Available in multiple small pack sizes for screening and synthesis.
  • Supplied as a solid, light brown to brown, stable under recommended storage.
  • Analytical documentation available: data sheet, COA, HNMR, LCMS, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.