Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(4-methoxyphenyl)pyridazin-3-amine | 4776-87-8 | MFCD06801353 | 97.3% | 201.23 g/mol | C11H11N3O | 100 MG
SDP

Catalog No. 5000300459
Encompass_Preferred

6-(4-methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic intermediate, notably in the preparation of SR 95103, a selective and competitive GABA-A receptor antagonist. It is supplied as a solid with reported high purity and established storage recommendations for both powder and solution forms.

  • High reported purity (97.31%).
  • Molecular formula C11H11N3O; molecular weight 201.23 g/mol.
  • Solid appearance suitable for handling and storage.
  • Recommended storage: powder at -20°C (long-term) or 4°C; in solvent at -80°C (short-term).
  • Available in small-scale packaging for research use.
  • Suitable as an intermediate for medicinal chemistry and receptor ligand synthesis.

Catalog No. 50-003-00459 Supplier Medchemexpress LLC Supplier No. HYW025443100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-(4-methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic intermediate, notably in the preparation of SR 95103, a selective and competitive GABA-A receptor antagonist. It is supplied as a solid with reported high purity and established storage recommendations for both powder and solution forms.

  • High reported purity (97.31%).
  • Molecular formula C11H11N3O; molecular weight 201.23 g/mol.
  • Solid appearance suitable for handling and storage.
  • Recommended storage: powder at -20°C (long-term) or 4°C; in solvent at -80°C (short-term).
  • Available in small-scale packaging for research use.
  • Suitable as an intermediate for medicinal chemistry and receptor ligand synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.