Learn More
Medchemexpress LLC 6-(4-methoxyphenyl)pyridazin-3-amine | 4776-87-8 | MFCD06801353 | 97.3% | 201.23 g/mol | C11H11N3O | 5 MG
SDP
Catalog No. 5000301249
Encompass_Preferred
6-(4-Methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic intermediate (for example in the synthesis of SR 95103). It is supplied as a light brown solid with reported purity of 97.31%, molecular weight 201.23 g/mol, and formula C11H11N3O. Soluble in DMSO and provided with supporting COA and SDS for research use.
  • High purity suitable for synthetic work.
  • Light brown solid, easy to handle.
  • Soluble in DMSO (≥ 15 mg/mL).
  • Provides long-term powder stability at -20°C.
  • Accompanied by certificate of analysis and safety data sheet.
  • Useful as an intermediate in receptor antagonist synthesis.
Catalog No. 50-003-01249 Supplier Medchemexpress LLC Supplier No. HYW0254435MG
6-(4-Methoxyphenyl)pyridazin-3-amine is an aminopyridazine derivative used as a synthetic intermediate (for example in the synthesis of SR 95103). It is supplied as a light brown solid with reported purity of 97.31%, molecular weight 201.23 g/mol, and formula C11H11N3O. Soluble in DMSO and provided with supporting COA and SDS for research use.
  • High purity suitable for synthetic work.
  • Light brown solid, easy to handle.
  • Soluble in DMSO (≥ 15 mg/mL).
  • Provides long-term powder stability at -20°C.
  • Accompanied by certificate of analysis and safety data sheet.
  • Useful as an intermediate in receptor antagonist synthesis.