Learn More
Medchemexpress LLC 6-(4-methylpiperazin-1-yl)-3-naphthalen-2-yl-4-pyridin-4-ylpyridazine | 1628502-91-9 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12011110MM 1ML
Encompass_Preferred
MW-150 is a selective, CNS-penetrant inhibitor of p38α mitogen-activated protein kinase (p38α MAPK). Supplied as a 10 mM solution in DMSO (1 mL), it has formula C24H23N5, molecular weight 381.47 g/mol, and high purity suitable for research applications.
  • Selective inhibition of p38α MAPK.
  • CNS penetrant and orally active in preclinical studies.
  • High purity (99.9%) suitable for research.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • Commonly used in neuroscience and inflammation research.
Catalog No. 50-002-33900