Learn More
Electron Microscopy Sciences Glass Knife Boat 6.4 mm
SDP
Catalog No. 50930662
Encompass_Preferred
These disposable plastic boats which easily attach to the triangular glass knife making a trough. To use simply slip the boat on the glass knife back and seal with either hot melted wax or nail polish.
Catalog No. 50-930-662 Supplier Electron Microscopy Sciences Supplier No. 71008
These disposable plastic boats which easily attach to the triangular glass knife making a trough. To use simply slip the boat on the glass knife back and seal with either hot melted wax or nail polish.