Learn More
Sigma Aldrich 6-(4-Phenylpiperazin-1-yl)pyridin-3-amine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1,1,1-trifluoro-2-propanol, a fluoroalcohol, is an organic solvent
- Vibrational spectral analysis of oh and od groups of 1,1,1-trifluoro-2-propanol and its deuterated derivatives has been conducted.1,1,1-trifluoro-2-propanol may be used in the synthesis of 1,1,1-trifluoropropan-2-yl 2-(4-hydroxy-3-methoxyphenyl)acetate and 1,1,1-trifluoropropan-2-yl 2-(3-ethoxy-4-hydroxyphenyl)acetate
Specifications
Specifications
| CAS | 927989-62-6 |
| Molecular Formula | C15H18N4 |
| Linear Formula | C15H18N4 |
| MDL Number | MFCD08165798 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 254.33 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.