Learn More
Medchemexpress LLC 6-[4-(trifluoromethoxy)phenyl]-3-(trifluoromethyl)-1,2,4-triazolo[4,3-a]pyridine | 1262618-39-2 | 99.6% | 347.22 | C14H7F6N3O | 200MG
SDP
Catalog No. 5000239161
Encompass_Preferred
GS967 (GS-458967) is a potent, selective inhibitor of the cardiac late sodium current (late INa) used as a research tool in preclinical electrophysiology and arrhythmia studies. It preferentially blocks persistent NaV1.5 currents and has reported IC50 values of 0.13 μM in ventricular myocytes and 0.21 μM in isolated hearts.
  • Potent, selective late INa inhibition suitable for arrhythmia research.
  • Reported IC50: 0.13 μM (ventricular myocytes), 0.21 μM (isolated hearts).
  • High purity (approximately 99.6%), with certificate of analysis available.
  • Molecular formula C14H7F6N3O; molecular weight 347.22 g/mol.
  • Supplied as a powder; recommended storage -20°C for long-term storage, -80°C for solutions.
  • Available in multiple package sizes to support assay development and in vivo studies.
Catalog No. 50-002-39161 Supplier Medchemexpress LLC Supplier No. HY12593200MG
GS967 (GS-458967) is a potent, selective inhibitor of the cardiac late sodium current (late INa) used as a research tool in preclinical electrophysiology and arrhythmia studies. It preferentially blocks persistent NaV1.5 currents and has reported IC50 values of 0.13 μM in ventricular myocytes and 0.21 μM in isolated hearts.
  • Potent, selective late INa inhibition suitable for arrhythmia research.
  • Reported IC50: 0.13 μM (ventricular myocytes), 0.21 μM (isolated hearts).
  • High purity (approximately 99.6%), with certificate of analysis available.
  • Molecular formula C14H7F6N3O; molecular weight 347.22 g/mol.
  • Supplied as a powder; recommended storage -20°C for long-term storage, -80°C for solutions.
  • Available in multiple package sizes to support assay development and in vivo studies.