Learn More
Cayman Chemical 6-(4-Methoxyphenyl)-3-pyridazinamine, Purity: ≥98%, CAS: 4776-87-8, 10 mg
SDP
Catalog No. 502965768
Encompass_Preferred
An aminopyridazine derivative that acts as a GABAA receptor antagonist that can be used as an intermediate in the synthesis of SR 95531
Catalog No. 50-296-5768 Supplier Cayman Chemical Supplier No. 1758810 MG
An aminopyridazine derivative that acts as a GABAA receptor antagonist that can be used as an intermediate in the synthesis of SR 95531