Learn More
Medchemexpress LLC 6-(5-chloropyridin-2-yl)-6,7-dihydro-5h-pyrrolo[3,4-b]pyrazin-5-one | 148891-53-6 | MFCD27967008 | 99.8% | 246.65 | C11H7ClN4O | 5 MG
SDP
Catalog No. 5000223427
Encompass_Preferred
RP 48497 is a photodegradation impurity of eszopiclone provided as a high-purity analytical reference for research, method development, and impurity profiling. It is characterized by a defined molecular formula, validated purity, and recommended storage conditions for stability.
Catalog No. 50-002-23427 Supplier Medchemexpress LLC Supplier No. HY1353595MG
RP 48497 is a photodegradation impurity of eszopiclone provided as a high-purity analytical reference for research, method development, and impurity profiling. It is characterized by a defined molecular formula, validated purity, and recommended storage conditions for stability.