Learn More
Medchemexpress LLC 6-(5-((MeSO2Et-amino)methyl)furan-2-yl)-N-(3-methyl-4-triazolopyridin-7-yloxyphenyl)quinazolin-4-amine | 937265-83-3 | MFCD22420817 | 99.2% | 569.63 g/mol | C29H27N7O4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1053110MM 1ML
Encompass_Preferred
ARRY-380 analog is a research small molecule provided as a ready-to-use solution for cellular and biochemical studies. It has been characterized in cell-based assays for effects on apoptotic pathways and intracellular pathogen replication.
  • Supplied as a 10 mM solution in DMSO for ready-to-use experiments.
  • Reported to activate caspase-3 and caspase-8 in cell-based studies.
  • Shown to inhibit Salmonella replication in cellular assays.
  • High reported purity (~99.2%) suitable for research applications.
  • Molecular weight 569.63 g/mol and formula C29H27N7O4S.
  • MFCD22420817 and CAS 937265-83-3 available for database cross-reference.
Catalog No. 50-002-43479