Learn More
Medchemexpress LLC 6-(5-(methoxymethyl)pyridin-3-yl)-3,4-dihydro-1,8-naphthyridine-1(2H)-carboxamide | 1633009-87-6 | MFCD28502113 | 99.2% | 298.34 g/mol | C16H18N4O2 | 10 MG
SDP
Catalog No. 5000212425
Encompass_Preferred
BI 689648 is a novel, highly selective aldosterone synthase (CYP11B2) inhibitor characterized for research use. It potently inhibits CYP11B2 (reported IC50 2.1 nM) while showing substantially reduced activity against CYP11B1 (IC50 310 nM), and has been profiled in nonhuman primate pharmacology studies. The compound is supplied as a purified research chemical for biochemical and in vivo investigations.
  • Potent and selective inhibition of aldosterone synthase (CYP11B2).
  • Reduced activity against cortisol synthase (CYP11B1) compared with CYP11B2.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Supplied as a high-purity research compound.
  • Documented chemical identifiers (CAS 1633009-87-6, formula C16H18N4O2).
Catalog No. 50-002-12425 Supplier Medchemexpress LLC Supplier No. HY10121710MG
BI 689648 is a novel, highly selective aldosterone synthase (CYP11B2) inhibitor characterized for research use. It potently inhibits CYP11B2 (reported IC50 2.1 nM) while showing substantially reduced activity against CYP11B1 (IC50 310 nM), and has been profiled in nonhuman primate pharmacology studies. The compound is supplied as a purified research chemical for biochemical and in vivo investigations.
  • Potent and selective inhibition of aldosterone synthase (CYP11B2).
  • Reduced activity against cortisol synthase (CYP11B1) compared with CYP11B2.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Supplied as a high-purity research compound.
  • Documented chemical identifiers (CAS 1633009-87-6, formula C16H18N4O2).