Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(5-methoxymethyl-pyridin-3-yl)-3,4-dihydro-2H-naphthyridine-1-carboxylic acid amide | 1633009-87-6 | 99.2% | 298.34 | C16H18N4O2 | 25 MG
SDP

Catalog No. 5000211108
Encompass_Preferred

The compound is a small-molecule, highly selective aldosterone synthase (CYP11B2) inhibitor with nanomolar potency, used in biochemical and pharmacological research to probe aldosterone synthesis and CYP11B2-mediated pathways. It exhibits reported IC50 values of 2.1 nM for CYP11B2 and 310 nM for CYP11B1, and is supplied as a high-purity solid for laboratory studies.

  • Highly selective aldosterone synthase inhibitor with nanomolar potency (CYP11B2 IC50 ≈ 2.1 nM).
  • Reduced off-target activity against CYP11B1 (IC50 ≈ 310 nM).
  • Molecular formula C16H18N4O2; molecular weight 298.34 g/mol.
  • Purity 99.2% as reported by supplier.
  • Supplied as a small solid quantity suitable for in vitro and in vivo research (e.g., 25 mg).
  • Characterized in peer-reviewed pharmacology studies for selectivity and pharmacodynamics.

Catalog No. 50-002-11108 Supplier Medchemexpress LLC Supplier No. HY10121725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The compound is a small-molecule, highly selective aldosterone synthase (CYP11B2) inhibitor with nanomolar potency, used in biochemical and pharmacological research to probe aldosterone synthesis and CYP11B2-mediated pathways. It exhibits reported IC50 values of 2.1 nM for CYP11B2 and 310 nM for CYP11B1, and is supplied as a high-purity solid for laboratory studies.

  • Highly selective aldosterone synthase inhibitor with nanomolar potency (CYP11B2 IC50 ≈ 2.1 nM).
  • Reduced off-target activity against CYP11B1 (IC50 ≈ 310 nM).
  • Molecular formula C16H18N4O2; molecular weight 298.34 g/mol.
  • Purity 99.2% as reported by supplier.
  • Supplied as a small solid quantity suitable for in vitro and in vivo research (e.g., 25 mg).
  • Characterized in peer-reviewed pharmacology studies for selectivity and pharmacodynamics.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.