Learn More
Roboz Surgical 6.5IN SURGICAL SCISSORS
SDP
Catalog No. NC9435044
Encompass
Kelly Scissors; Straight; Sharp; 29mm Blade Length; 6-1/4 in. Overall Length
Catalog No. NC9435044 Supplier Roboz Surgical Supplier No. RS6050
Kelly Scissors; Straight; Sharp; 29mm Blade Length; 6-1/4 in. Overall Length