Learn More
Strem, An Ascensus Company CAS# 121627-17-6. 250mg. 6,6'-[(3,3'-Di-t-butyl-5,5'-dimethoxy-1,1'-biphenyl-2,2'-diyl)bis(oxy)] bis(dibenzo[d,f][1,3,2]dioxaphosphepin) hemi ethyl acetate adduct, min. 95% BIPHEPHOS. MFCD09839135
SDP
Supplier:  Strem, An Ascensus Company 150107250MG
Encompass
CAS# 121627-17-6. 250mg. 6,6'-[(3,3'-Di-t-butyl-5,5'-dimethoxy-1,1'-biphenyl-2,2'-diyl)bis(oxy)] bis(dibenzo[d,f][1,3,2]dioxaphosphepin) hemi ethyl acetate adduct, min. 95% BIPHEPHOS. MFCD09839135. Molecular Weight: 786.78 (830.84). Molecular Formula: C46H44O8P2·;0.5EtOAc. Color/form: white to off-white pwdr. Strem# 15-0107. http://www.strem.com/catalog/v/15-0107/
Catalog No. 50-901-09746