Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(6-(4-hydroxypiperidinyl)hexyloxy)-3-methylflavone hydrochloride | 139652-86-1 | 99.1% | 472.02 g·mol⁻¹ | C27H34ClNO4 | 1 ML
SDP

Catalog No. 5000246051
Encompass_Preferred

Sigma-LIGAND-1 hydrochloride is a selective sigma receptor ligand supplied as a hydrochloride salt for use in receptor binding and pharmacology research. It is provided both as a high-purity solid and as a ready-to-use solution in DMSO, with documented solubility and storage recommendations for reliable experimental use.

  • Selective sigma receptor ligand for binding and pharmacology studies.
  • High purity (≈99.1%) for consistent assay performance.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Off-white to light yellow solid available in multiple sizes.
  • Soluble in DMSO with ultrasonic warming; use newly opened DMSO.
  • Recommended storage: solid refrigerated for long-term; solutions frozen for stability.

Catalog No. 50-002-46051 Supplier Medchemexpress LLC Supplier No. HY101626A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sigma-LIGAND-1 hydrochloride is a selective sigma receptor ligand supplied as a hydrochloride salt for use in receptor binding and pharmacology research. It is provided both as a high-purity solid and as a ready-to-use solution in DMSO, with documented solubility and storage recommendations for reliable experimental use.

  • Selective sigma receptor ligand for binding and pharmacology studies.
  • High purity (≈99.1%) for consistent assay performance.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Off-white to light yellow solid available in multiple sizes.
  • Soluble in DMSO with ultrasonic warming; use newly opened DMSO.
  • Recommended storage: solid refrigerated for long-term; solutions frozen for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.