Learn More
Medchemexpress LLC 6-((6,7-Bis(methoxy-d3)quinazolin-4-yl)oxy)-N,2-dimethylbenzofuran-3-carboxamide | 2978508-76-6 | 99.5% | 399.43 | 1 MG
SDP
Catalog No. 5000401819
Encompass_Preferred
Fruquintinib-d6 is a deuterium-labeled analog of Fruquintinib, a selective and potent oral inhibitor of VEGFR-1, -2, and -3. This product is intended for research use only and has not been fully validated for medical applications. It is supplied as a white to off-white solid powder and should be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities. This product is classified as hazardous with several precautionary statements.
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Stable under recommended storage conditions
Catalog No. 50-004-01819 Supplier Medchemexpress LLC Supplier No. HY19912S1MG
Fruquintinib-d6 is a deuterium-labeled analog of Fruquintinib, a selective and potent oral inhibitor of VEGFR-1, -2, and -3. This product is intended for research use only and has not been fully validated for medical applications. It is supplied as a white to off-white solid powder and should be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities. This product is classified as hazardous with several precautionary statements.
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
  • Stable under recommended storage conditions