Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6'-Bis(chloromethyl)-2,2'-bipyridyl 98.0+%, TCI America™
SDP

Catalog No. B4645200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4645200MG 200 mg
B46451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4645200MG Supplier TCI America Supplier No. B4645200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 74065-64-8 | C12H10Cl2N2 | 253.13 g/mol

Chemical Identifiers

CAS 74065-64-8
Molecular Formula C12H10Cl2N2
Molecular Weight (g/mol) 253.13
MDL Number MFCD02181082
InChI Key CUWSIBMCPYEBPL-UHFFFAOYSA-N
Synonym 6,6′C-Bis(chloromethyl)-2,2′C-bipyridine
PubChem CID 1502060
IUPAC Name 6,6'-bis(chloromethyl)-2,2'-bipyridine
SMILES ClCC1=CC=CC(=N1)C1=NC(CCl)=CC=C1

Specifications

Melting Point 162°C
Color White
Quantity 200 mg
UN Number 3261
Formula Weight 253.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′-Bis(chloromethyl)-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.