Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-6,6'-Dibromo-1,1'-bi-2-naphthol 98.0+%, TCI America™
SDP

Catalog No. D27791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27791G 1 g
D27795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27791G Supplier TCI America Supplier No. D27791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13185-00-7
Molecular Formula C20H12Br2O2
Molecular Weight (g/mol) 444.122
MDL Number MFCD00798290
InChI Key OORIFUHRGQKYEV-UHFFFAOYSA-N
Synonym r---6,6'-dibromo-1,1'-bi-2-naphthol, s---6,6'-dibromo-1,1'-bi-2-naphthol, s-+-6,6'-dibromo-1,1'-bi-2-naphthol, 6,6'-dibromo-1,1'-bi-2-naphthol, s-+-6,6'-dibromo-1,1'-binaphthalene-2,2'-diol, r---6,6'-dibromo-1,1'-binaphthalene-2,2'-diol, 6,6'-dibromo 1,1'-binaphthalene-2,2'-diol, +/--6,6'-dibromo-1,1'-bi-2-naphthol, s-6,6'-dibromo-1,1'-bi-2-naphthol, racemic-6,6'-dibromo-1,1'-bi-2-naphthol
PubChem CID 222842
IUPAC Name 6-bromo-1-(6-bromo-2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2C3=C(C=CC4=C3C=CC(=C4)Br)O)O)C=C1Br

Specifications

Melting Point 205°C
Color White-Yellow
Quantity 1 g
Formula Weight 444.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-6,6′-Dibromo-1,1′-bi-2-naphthol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.