Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6'-Dimethyl-2,2'-dipyridyl, Thermo Scientific™

Catalog No. AC469310050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469310050 5 g
AC469310010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469310050 Supplier Thermo Scientific Chemicals Supplier No. 469310050
Add to Cart
This item is not returnable. View return policy

Specifications

Alpha Vector DIMETHYLDIPYRIDYL
Chemical Name or Material 6,6 '-Dimethyl-2,2 '-dipyridyl
CAS 4411-80-7
Melting Point 86°C to 96°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C12 H12 N2
Quantity 5 g
MDL Number 00059776
Synonym 6,6 '-Bi-2-picoline
Molecular Weight (g/mol) 184.24
Physical Form Crystalline Powder and/or Chunks
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.