Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [6.6] Diphenyl C62 bis(butyric acid methyl ester)(mixture of isomers)

Catalog No. 704326100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-432-6100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-432-6100MG Supplier Sigma Aldrich Supplier No. 704326100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[6.6] Diphenyl C62 bis(butyric acid methyl ester)(mixture of isomers)

Specifications

Melting Point >300°C
Molecular Formula C84H28O4
Linear Formula C84H28O4
Quantity 100 mg
Synonym Bis(1-[3-(methoxycarbonyl)propyl]-1-phenyl)-[6.6 ]C62 (mixture of isomers); Bis[60 ]PCBM
Molecular Weight (g/mol) 1101.12
Percent Purity 99.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.