Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [6,6]-Phenyl C61 butyric acid methyl ester

Catalog No. 684457100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-445-7100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-445-7100MG Supplier Sigma Aldrich Supplier No. 684457100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[6,6]-Phenyl C61 butyric acid methyl ester ([60]PCBM) is a methanofullerene that has a better solubility in organic solvents than fullerenes(C60). It has high electron mobility which enables its function as an electron acceptor in electrochemical applications.[60]PCBM is an n-type semi-conductor widely used as an electron transport material with low cost and high surface area in different energy based applications like organic photovoltaics (OPVs), perovskite solar cells (PSCs), field effect transistors (FETs) and photodetectors.

Specifications

CAS 160848-22-6
Molecular Formula C72H14O2
Linear Formula C72H14O2
Quantity 100 mg
MDL Number MFCD07784544
Synonym 1-[3-(Methoxycarbonyl)propyl]-1-phenyl-[6.6 ]C61; 3 H-Cyclopropa[1,9 ] [5,6 ]fullerene-C60-Ih-3 -butanoic acid 3 -phenyl methyl ester; PCBM; [60 ]PCBM
Molecular Weight (g/mol) 910.88
Percent Purity >99.9%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.