Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [6,6]-Phenyl C61 butyric acid methyl ester

Catalog No. 684449500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-444-9500MG 500 mg
68-444-9100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68-444-9500MG Supplier Sigma Aldrich Supplier No. 684449500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[6,6]-Phenyl C61butyric acid methyl ester ([60]PCBM) is a methanofullerene that has a better diffusion in organic molecules than fullerenes (C60). It has high electron mobility, which allows it to be used as an electron acceptor in major electrochemical applications.[60]PCBM is an n-type semi-conductor with low cost and high surface area. It is widely used as an electron transport material in various energy-based applications like organic photovoltaics, perovskite solar cells, field effect transistors, and photodetectors.

Specifications

CAS 160848-22-6
Molecular Formula C72H14O2
Linear Formula C72H14O2
MDL Number MFCD07784544
Quantity 500 mg
Synonym 1-[3-(Methoxycarbonyl)propyl]-1-phenyl-[6.6 ]C61; 3 H-Cyclopropa[1,9 ] [5,6 ]fullerene-C60-Ih-3 -butanoic acid 3 -phenyl methyl ester; PCBM; [60 ]PCBM
Molecular Weight (g/mol) 910.88
Percent Purity >99.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.