Learn More
Cayman Chemical 6,6′-Trehalose Dioleate, Purity: ≥95%, CAS: 338733-40-7, 5 mg
SDP
Catalog No. 502945010
Encompass_Preferred
A glycolipid; has been used in the generation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing 6,6′-TDO and encapsulating mRNA encoding influenza HA increase Ifn-γ, Il-2, and Tnf-α secretion by isolated mouse splenocytes; LNPs containing 6,6′-TDO accumulate in the liver and spleen after intramuscular administration in mice; unlike LNP formulations that do not contain 6,6′-TDO, LNPs containing 6,6′-TDO do not decrease serum levels of BUN or increase serum levels of LDH, troponin I, ALT, and AST in mice; LNPs containing 6,6′-TDO and encapsulating mRNA encoding influenza A HA increase serum IgG1 and IgG2a titers in mice
Catalog No. 50-294-5010 Supplier Cayman Chemical Supplier No. 422125 MG
A glycolipid; has been used in the generation of LNPs for the delivery of mRNA in vitro and in vivo; LNPs containing 6,6′-TDO and encapsulating mRNA encoding influenza HA increase Ifn-γ, Il-2, and Tnf-α secretion by isolated mouse splenocytes; LNPs containing 6,6′-TDO accumulate in the liver and spleen after intramuscular administration in mice; unlike LNP formulations that do not contain 6,6′-TDO, LNPs containing 6,6′-TDO do not decrease serum levels of BUN or increase serum levels of LDH, troponin I, ALT, and AST in mice; LNPs containing 6,6′-TDO and encapsulating mRNA encoding influenza A HA increase serum IgG1 and IgG2a titers in mice