Learn More
Cayman Chemical 6 6DImethyl3 4methylbenzo 10mg
SDP
Catalog No. 502943386
Encompass_Preferred
A GLP-1R positive allosteric modulator; increases GLP-1-induced cAMP production by 268.35% in a reporter assay using HEK293 cells expressing human GLP-1R; induces insulin secretion in glucose-stimulated GLP-1 (7-36)-induced INS-1 832/13 cells at 5 µM; decreases blood glucose levels in a glucose tolerance test in fasted mice at 10 or 30 mg/kg
Catalog No. 50-294-3386 Supplier Cayman Chemical Supplier No. 4217510 MG
A GLP-1R positive allosteric modulator; increases GLP-1-induced cAMP production by 268.35% in a reporter assay using HEK293 cells expressing human GLP-1R; induces insulin secretion in glucose-stimulated GLP-1 (7-36)-induced INS-1 832/13 cells at 5 µM; decreases blood glucose levels in a glucose tolerance test in fasted mice at 10 or 30 mg/kg