Learn More
Cayman Chemical 6 7 4-TrIhydroxyIsoflavon 10mg
SDP
Catalog No. 502913789
Encompass_Preferred
An active metabolite of daidzein; suppresses anchorage-dependent and -independent growth of HCT116 and DLD-1 colon cancer cells, as well as induces cell cycle arrest at the S and G2/M phases in HCT116 cells from 12.5-100 µM; inhibits MDI-induced adipogenesis in 3T3-L1 preadipocytes at 40 and 80 µM; reverses scopolamine-induced memory impairments and increases hippocampal BDNF and CREB levels in mice at 5 mg/kg; prevents LPS-induced bone loss in mice
Catalog No. 50-291-3789 Supplier Cayman Chemical Supplier No. 3540510 MG
An active metabolite of daidzein; suppresses anchorage-dependent and -independent growth of HCT116 and DLD-1 colon cancer cells, as well as induces cell cycle arrest at the S and G2/M phases in HCT116 cells from 12.5-100 µM; inhibits MDI-induced adipogenesis in 3T3-L1 preadipocytes at 40 and 80 µM; reverses scopolamine-induced memory impairments and increases hippocampal BDNF and CREB levels in mice at 5 mg/kg; prevents LPS-induced bone loss in mice