Learn More
Cayman Chemical 6,7,4'-Trihydroxyisoflavone, Purity: ≥98%, CAS: 17817-31-1, 100 mg
SDP
Catalog No. 502909405
Encompass_Preferred
An active metabolite of daidzein; suppresses anchorage-dependent and -independent growth of HCT116 and DLD-1 colon cancer cells, as well as induces cell cycle arrest at the S and G2/M phases in HCT116 cells from 12.5-100 µM; inhibits MDI-induced adipogenesis in 3T3-L1 preadipocytes at 40 and 80 µM; reverses scopolamine-induced memory impairments and increases hippocampal BDNF and CREB levels in mice at 5 mg/kg; prevents LPS-induced bone loss in mice
Catalog No. 50-290-9405 Supplier Cayman Chemical Supplier No. 35405100 MG
An active metabolite of daidzein; suppresses anchorage-dependent and -independent growth of HCT116 and DLD-1 colon cancer cells, as well as induces cell cycle arrest at the S and G2/M phases in HCT116 cells from 12.5-100 µM; inhibits MDI-induced adipogenesis in 3T3-L1 preadipocytes at 40 and 80 µM; reverses scopolamine-induced memory impairments and increases hippocampal BDNF and CREB levels in mice at 5 mg/kg; prevents LPS-induced bone loss in mice