Learn More
Medchemexpress LLC 6 7-Dichloroquinolin 10g
SDP
Catalog No. 5000205141
Encompass_Preferred
6 7-Dichloroquinoline-5 8-dione (compound DA3002) is a potent CDC25B2 inhibitor with an IC50 value of 4 6 M 6 7-Dichloroquinoline-5 8-dione has the potential for the research of cancer[1]
Catalog No. 50-002-05141 Supplier Medchemexpress LLC Supplier No. HYW10293110G
6 7-Dichloroquinoline-5 8-dione (compound DA3002) is a potent CDC25B2 inhibitor with an IC50 value of 4 6 M 6 7-Dichloroquinoline-5 8-dione has the potential for the research of cancer[1]