Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6,7-diethoxy-4-methylcoumarin | 314744-06-4 | 98.0% | 248.27 | C14H16O4 | 10 MG
SDP

Catalog No. 5000297321
Encompass_Preferred

6,7-Diethoxy-4-methylcoumarin is a coumarin derivative provided as a fluorogenic substrate for biochemical and analytical assays, and is reported as a metabolite of Cordyceps militaris. It exhibits excitation near 344 nm and emission near 445 nm, and is supplied as a light brown solid with high purity for research use.

  • Fluorogenic substrate suitable for fluorescence-based enzyme and metabolic assays.
  • Excitation ~344 nm and emission ~445 nm for fluorescence detection.
  • High purity (98.0%) for analytical and research applications.
  • Solid, light brown physical form convenient for handling and storage.
  • Molecular formula C14H16O4 and molecular weight 248.27 g/mol.
  • Supplied in small-mass quantities for analytical and probe use.

Catalog No. 50-002-97321 Supplier Medchemexpress LLC Supplier No. HYD006310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6,7-Diethoxy-4-methylcoumarin is a coumarin derivative provided as a fluorogenic substrate for biochemical and analytical assays, and is reported as a metabolite of Cordyceps militaris. It exhibits excitation near 344 nm and emission near 445 nm, and is supplied as a light brown solid with high purity for research use.

  • Fluorogenic substrate suitable for fluorescence-based enzyme and metabolic assays.
  • Excitation ~344 nm and emission ~445 nm for fluorescence detection.
  • High purity (98.0%) for analytical and research applications.
  • Solid, light brown physical form convenient for handling and storage.
  • Molecular formula C14H16O4 and molecular weight 248.27 g/mol.
  • Supplied in small-mass quantities for analytical and probe use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.