Learn More
AdipoGen 6 7-Diethoxy-4-methylcoumarin
SDP
Supplier:  AdipoGen CDXD0028M100
Encompass_Preferred
Chemical. CAS 314744-06-4. Formula C14H16O4. MW 248.27. Synthetic. Fluorogenic dealkylase substrate for the measurement of the cytochrome-mediated monooxygenase or "mixed function" oxidase system.
Catalog No. 50-256-1306