Learn More
Medchemexpress LLC 6,7-Dihydro-4-benzo[b]thiophenone | 13414-95-4 | 99.5% | C8H8OS | 1 ML
SDP
Catalog No. 5000434934
Encompass_Preferred
6,7-Dihydro-4-benzo[b]thiophenone is a biochemical reagent designed for various life science-related research applications. It serves as a versatile biological material or an organic compound for laboratory use.
  • Suitable as a biological material or organic compound
  • Intended for research use only
  • Soluble in DMSO at 100 mg/mL
  • Pure form stable at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-34934 Supplier Medchemexpress LLC Supplier No. HY7866710MM/1ML
6,7-Dihydro-4-benzo[b]thiophenone is a biochemical reagent designed for various life science-related research applications. It serves as a versatile biological material or an organic compound for laboratory use.
  • Suitable as a biological material or organic compound
  • Intended for research use only
  • Soluble in DMSO at 100 mg/mL
  • Pure form stable at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month