Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Dihydroxycoumarin-3-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. D41705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41705G 5 g
D41701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41705G Supplier TCI America Supplier No. D41705G

Chemical Identifiers

CAS 84738-35-2
Molecular Formula C10H6O6
Molecular Weight (g/mol) 222.15
MDL Number MFCD18450959
InChI Key STCWBNGIVIFZHI-UHFFFAOYSA-N
PubChem CID 13989553
IUPAC Name 6,7-dihydroxy-2-oxo-2H-chromene-3-carboxylic acid
SMILES OC(=O)C1=CC2=CC(O)=C(O)C=C2OC1=O

Specifications

Melting Point 270°C
Quantity 5 g
Formula Weight 222.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dihydroxycoumarin-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.