Learn More
Apexbio Technology LLC 6,7-Dihydroxycoumarin 305-01-1 20mg
SDP
Catalog No. 5000087710
Encompass_Preferred
6 7-Dihydroxycoumarin (CAS 305-01-1) also known as esculetin is a naturally occurring coumarin derivative extracted from Aesculus hippocastanum Linn This compound exhibits notable antioxidant activity primarily by scavenging reactive oxygen species and inhibiting lipid peroxidation Additionally 6 7-dihydroxycoumarin has been studied for its modulatory effects on various cellular signaling pathways implicated in inflammation and apoptosis Its applications in biomedical research include investigations into oxidative stress inflammation and related disease models The compound is soluble in ethanol and dilute alkali and should be stored in a sealed cool and dry environment to preserve stability
Catalog No. 50-000-87710 Supplier Apexbio Technology LLC Supplier No. N176420
6 7-Dihydroxycoumarin (CAS 305-01-1) also known as esculetin is a naturally occurring coumarin derivative extracted from Aesculus hippocastanum Linn This compound exhibits notable antioxidant activity primarily by scavenging reactive oxygen species and inhibiting lipid peroxidation Additionally 6 7-dihydroxycoumarin has been studied for its modulatory effects on various cellular signaling pathways implicated in inflammation and apoptosis Its applications in biomedical research include investigations into oxidative stress inflammation and related disease models The compound is soluble in ethanol and dilute alkali and should be stored in a sealed cool and dry environment to preserve stability