Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6,7-Dimethoxy-2-(2-phenylethyl)chromone | 84294-87-1 | 1 MG
SDP

Catalog No. 5000417659
Encompass_Preferred

6,7-Dimethoxy-2-(2-phenylethyl)chromone is a natural compound intended for research use only. It has a molecular weight of 310.34 and a chemical formula of C19H18O4. This compound is a solid, appearing white to off-white.

  • Natural compound for research use
  • Purity 98.99%
  • Molecular weight: 310.34
  • Chemical formula: C19H18O4
  • Appearance: Solid
  • Color: White to off-white
  • Initial source from plants
  • Solid storage: 4°C, sealed, away from moisture
  • Solution storage: -80°C for 6 months, or -20°C for 1 month, sealed, away from moisture

Catalog No. 50-004-17659 Supplier Medchemexpress LLC Supplier No. HYN81231MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6,7-Dimethoxy-2-(2-phenylethyl)chromone is a natural compound intended for research use only. It has a molecular weight of 310.34 and a chemical formula of C19H18O4. This compound is a solid, appearing white to off-white.

  • Natural compound for research use
  • Purity 98.99%
  • Molecular weight: 310.34
  • Chemical formula: C19H18O4
  • Appearance: Solid
  • Color: White to off-white
  • Initial source from plants
  • Solid storage: 4°C, sealed, away from moisture
  • Solution storage: -80°C for 6 months, or -20°C for 1 month, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.