Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6,7-Dimethoxy-2-(2-phenylethyl)chromone | 84294-87-1 | 5 MG
SDP

Catalog No. 5000417555
Encompass_Preferred

6,7-Dimethoxy-2-(2-phenylethyl)chromone is a natural compound for research use only.

  • Not sold to patients
  • Purity 98.99%
  • Molecular weight 310.34
  • Molecular formula C19H18O4
  • Appearance solid
  • Color white to off-white
  • Stable under recommended storage conditions

Catalog No. 50-004-17555 Supplier Medchemexpress LLC Supplier No. HYN81235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6,7-Dimethoxy-2-(2-phenylethyl)chromone is a natural compound for research use only.

  • Not sold to patients
  • Purity 98.99%
  • Molecular weight 310.34
  • Molecular formula C19H18O4
  • Appearance solid
  • Color white to off-white
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.