Learn More
Medchemexpress LLC 6,7-dimethoxy-2-phenylquinoxaline | 146535-11-7 | MFCD00270913 | 99.7% | C16H14N2O2 | 10MG
SDP
Catalog No. 5000260164
Encompass_Preferred
Tyrphostin AG1296 is a small-molecule inhibitor of platelet-derived growth factor receptor (PDGFR) tyrosine kinase used in research to probe PDGFR-dependent signaling, cell viability, and migration. Supplied as a high-purity solid, it is provided with supporting QC and safety documents and is available in multiple small pack sizes.
  • Potent, selective PDGFR inhibitor for signaling studies
  • High purity suitable for biochemical and cell-based assays
  • Supplied as a solid powder for accurate weighing and dosing
  • Available in a range of small pack sizes for screening work
  • Stable storage profile for long-term powder preservation
  • Includes datasheet, certificate of analysis, and safety data sheet
Catalog No. 50-002-60164 Supplier Medchemexpress LLC Supplier No. HY1389410MG
Tyrphostin AG1296 is a small-molecule inhibitor of platelet-derived growth factor receptor (PDGFR) tyrosine kinase used in research to probe PDGFR-dependent signaling, cell viability, and migration. Supplied as a high-purity solid, it is provided with supporting QC and safety documents and is available in multiple small pack sizes.
  • Potent, selective PDGFR inhibitor for signaling studies
  • High purity suitable for biochemical and cell-based assays
  • Supplied as a solid powder for accurate weighing and dosing
  • Available in a range of small pack sizes for screening work
  • Stable storage profile for long-term powder preservation
  • Includes datasheet, certificate of analysis, and safety data sheet