Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Dimethoxy-3,4-dihydroisoquinoline 98.0+%, TCI America™
SDP

Catalog No. D50465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50465G 5 g
D50461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50465G Supplier TCI America Supplier No. D50465G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3382-18-1
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00266748
InChI Key NSLJVQUDZCZJLK-UHFFFAOYSA-N
PubChem CID 30058
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline
SMILES COC1=CC2=C(C=NCC2)C=C1OC

Specifications

Melting Point 49°C
Boiling Point 160°C
Quantity 5 g
Synonym 6,7-dimethoxy-3,4-dihydro-isoquinoline, 3,4-dihydro-6,7-dimethoxyisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy, pubchem12828, ksc571e8h, 3,4-dihydro-6,7-dimethoxy-isoquinoline, isoquinoline,3,4-dihydro-6,7-dimethoxy, isoquinoline, 3,4-dihydro-6,7-dimethoxy-, hydrochloride 1:1
Formula Weight 191.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 6,7-Dimethoxy-3,4-dihydroisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.