Learn More
Medchemexpress LLC 6,7-dimethoxy-N-(3,4,5-trimethoxyphenyl)quinolin-4-amine | 319492-82-5 | 98.8% | 370.405 | C20H22N2O5 | 50MG
SDP
Catalog No. 5000237036
Encompass_Preferred
GAK inhibitor 49 is a potent, ATP-competitive, and highly selective inhibitor of cyclin G-associated kinase (GAK) used as a research tool to probe GAK-related signaling. It exhibits sub-nanomolar binding affinity and low-nanomolar cellular potency, and is provided as a high-purity solid for in vitro studies.
  • Potent ATP-competitive inhibition of GAK (Ki = 0.54 nM).
  • High cellular potency suitable for cell-based assays (cell IC50 = 56 nM).
  • High purity for biochemical testing (>98%).
  • Well characterized molecular properties (MW 370.40 g/mol; C20H22N2O5).
  • Available in small research quantities for assay development and screening.
Catalog No. 50-002-37036 Supplier Medchemexpress LLC Supplier No. HY12479350MG
GAK inhibitor 49 is a potent, ATP-competitive, and highly selective inhibitor of cyclin G-associated kinase (GAK) used as a research tool to probe GAK-related signaling. It exhibits sub-nanomolar binding affinity and low-nanomolar cellular potency, and is provided as a high-purity solid for in vitro studies.
  • Potent ATP-competitive inhibition of GAK (Ki = 0.54 nM).
  • High cellular potency suitable for cell-based assays (cell IC50 = 56 nM).
  • High purity for biochemical testing (>98%).
  • Well characterized molecular properties (MW 370.40 g/mol; C20H22N2O5).
  • Available in small research quantities for assay development and screening.