Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6,7-dimethoxyflavone

Catalog No. CDS007277 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS007277 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS007277 Supplier Sigma Aldrich Supplier No. CDS00727710MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Thiomorpholine forms complexes with cu(ii), pt(ii) and ni(ii) salts and their catalytic activity has been investigated.thiomorpholine has been used in the preparation of:
  • N-boc-α-alkyl-α-(sec-amino)alanines
  • Thiomorpholine-n-borane

Specifications

Molecular Formula C17H14O4
Linear Formula C17H14O4
MDL Number MFCD00017698
Quantity 25 g
Molecular Weight (g/mol) 282.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.