Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6,7-diphenyl-2-sulfanylidene-1H-pteridin-4-one | 14892-97-8 | MFCD02167478 | 99.9% | 332.38 g/mol | C18H12N4OS | 50 MG
SDP

Catalog No. 5000242384
Encompass_Preferred

SCR7 pyrazine is a small-molecule inhibitor of DNA ligase IV that blocks non-homologous end-joining (NHEJ) and increases homology-directed repair (HDR) efficiency in genome-editing workflows. It is supplied as a light yellow solid with high analytical purity suitable for research applications.

  • Inhibits DNA ligase IV and reduces non-homologous end-joining.
  • Enhances CRISPR/Cas9-mediated homology-directed repair.
  • High purity (99.9%) for reliable experimental results.
  • Molecular weight 332.38 g/mol.
  • Solid, light yellow appearance for convenient handling.
  • Recommended storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (2 years).

Catalog No. 50-002-42384 Supplier Medchemexpress LLC Supplier No. HY10784550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SCR7 pyrazine is a small-molecule inhibitor of DNA ligase IV that blocks non-homologous end-joining (NHEJ) and increases homology-directed repair (HDR) efficiency in genome-editing workflows. It is supplied as a light yellow solid with high analytical purity suitable for research applications.

  • Inhibits DNA ligase IV and reduces non-homologous end-joining.
  • Enhances CRISPR/Cas9-mediated homology-directed repair.
  • High purity (99.9%) for reliable experimental results.
  • Molecular weight 332.38 g/mol.
  • Solid, light yellow appearance for convenient handling.
  • Recommended storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.