Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6,7-diphenyl-2-sulfanylidene-1H-pteridin-4-one | 14892-97-8 | MFCD02167478; MFCD28718010 | 99.9% | 332.38 g·mol⁻¹ | C18H12N4OS | 10 MG
SDP

Catalog No. 5000240315
Encompass_Preferred

SCR7 pyrazine is a small-molecule inhibitor of DNA ligase IV that blocks non-homologous end joining (NHEJ) and increases homology-directed repair (HDR) efficiency in genome-editing experiments. It is provided as a research-grade solid for use in in vitro and in vivo studies to modulate DNA repair pathways.

  • Inhibits DNA ligase IV and blocks NHEJ.
  • Enhances homology-directed repair efficiency in genome-editing workflows.
  • High chemical purity suitable for research applications.
  • Molecular formula C18H12N4OS and molecular weight 332.38 g·mol⁻¹.
  • Soluble in DMSO for preparation of concentrated stock solutions.

Catalog No. 50-002-40315 Supplier Medchemexpress LLC Supplier No. HY10784510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SCR7 pyrazine is a small-molecule inhibitor of DNA ligase IV that blocks non-homologous end joining (NHEJ) and increases homology-directed repair (HDR) efficiency in genome-editing experiments. It is provided as a research-grade solid for use in in vitro and in vivo studies to modulate DNA repair pathways.

  • Inhibits DNA ligase IV and blocks NHEJ.
  • Enhances homology-directed repair efficiency in genome-editing workflows.
  • High chemical purity suitable for research applications.
  • Molecular formula C18H12N4OS and molecular weight 332.38 g·mol⁻¹.
  • Soluble in DMSO for preparation of concentrated stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.