Learn More
Cayman Chemical 6 7-EpoxybergamotIn 10mg
SDP
Catalog No. 502984571
Encompass_Preferred
A potent CYP3A4 inhibitor (IC50s = 0.30 ppm in a cell-free assay, 0.33 µM in HL7 human liver cells, and 0.22 µM in S9 human intestine cells); inhibits the transport of talinolol, a P-gp/ABCB1 transporter substrate, across Caco-2 cell monolayers (IC50 = 0.7 µM)
Catalog No. 50-298-4571 Supplier Cayman Chemical Supplier No. 1976010 MG
A potent CYP3A4 inhibitor (IC50s = 0.30 ppm in a cell-free assay, 0.33 µM in HL7 human liver cells, and 0.22 µM in S9 human intestine cells); inhibits the transport of talinolol, a P-gp/ABCB1 transporter substrate, across Caco-2 cell monolayers (IC50 = 0.7 µM)