Learn More
Medchemexpress LLC 6,8,10,14-Eicosatetraenedioic acid, 5,12-dihydroxy-, (5S,6Z,8E,10E,12R,14Z)- | 80434-82-8 | 98.5% | 366.45 | 25 UG
SDP
Catalog No. 5000439924
Encompass_Preferred
20-Carboxy-Leukotriene B4 (20-COOH LTB4) is a metabolite of Leukotriene B4. It binds to the BLT1 receptor with high affinity and inhibits LTB4-mediated neutrophil responses, including migration, degranulation, and leukotriene biosynthesis.
  • Binds to the BLT1 receptor with high affinity.
  • Inhibits LTB4-mediated neutrophil responses.
  • It is a human endogenous metabolite.
  • Data sheet, COA, SDS, and handling instructions are available.
Catalog No. 50-004-39924 Supplier Medchemexpress LLC Supplier No. 25UGHY113483
20-Carboxy-Leukotriene B4 (20-COOH LTB4) is a metabolite of Leukotriene B4. It binds to the BLT1 receptor with high affinity and inhibits LTB4-mediated neutrophil responses, including migration, degranulation, and leukotriene biosynthesis.
  • Binds to the BLT1 receptor with high affinity.
  • Inhibits LTB4-mediated neutrophil responses.
  • It is a human endogenous metabolite.
  • Data sheet, COA, SDS, and handling instructions are available.