Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,8-Bis(benzylthio)octanoic Acid 98.0+%, TCI America™
SDP

Catalog No. B53685MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53685MG 5 mg
B536825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53685MG Supplier TCI America Supplier No. B53685MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95809-78-2
Molecular Formula C22H28O2S2
Molecular Weight (g/mol) 388.58
MDL Number MFCD22420826
InChI Key ZYRLHJIMTROTBO-UHFFFAOYNA-N
Synonym CPI 613
PubChem CID 24770514
IUPAC Name 6,8-bis(benzylsulfanyl)octanoic acid
SMILES OC(=O)CCCCC(CCSCC1=CC=CC=C1)SCC1=CC=CC=C1

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 5 mg
Formula Weight 388.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,8-Bis(benzylthio)octanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.