Learn More
Medchemexpress LLC 6,8-Diprenylorobol | 66777-70-6 | 1 MG
SDP
Catalog No. 5000421641
Encompass_Preferred
6,8-Diprenylorobol is a prenylated isoflavone isolated from the leaves of *Cudrania tricuspidata*. It exhibits an antiproliferative effect and induces apoptosis by activating p53 and generating reactive oxygen species (ROS). It has shown to inhibit proliferation in LoVo and HCT15 cells in a dose- and time-dependent manner and also regulates MAPKs.
  • Exhibits antiproliferative effect
  • Induces apoptosis by activating p53
  • Generates reactive oxygen species (ROS)
  • Inhibits proliferation in LoVo and HCT15 cells
  • Regulates MAPKs in LoVo and HCT15 cells
  • Increases ROS levels in LoVo and HCT15 cells
  • Inhibits LPS-induced nitric oxide production in RAW264.7 cells
Catalog No. 50-004-21641 Supplier Medchemexpress LLC Supplier No. HYN26931MG
6,8-Diprenylorobol is a prenylated isoflavone isolated from the leaves of *Cudrania tricuspidata*. It exhibits an antiproliferative effect and induces apoptosis by activating p53 and generating reactive oxygen species (ROS). It has shown to inhibit proliferation in LoVo and HCT15 cells in a dose- and time-dependent manner and also regulates MAPKs.
  • Exhibits antiproliferative effect
  • Induces apoptosis by activating p53
  • Generates reactive oxygen species (ROS)
  • Inhibits proliferation in LoVo and HCT15 cells
  • Regulates MAPKs in LoVo and HCT15 cells
  • Increases ROS levels in LoVo and HCT15 cells
  • Inhibits LPS-induced nitric oxide production in RAW264.7 cells