Learn More
Cayman Chemical 6 8DIfluoro7hydroxy4meth 10mg
SDP
Catalog No. 502911155
Encompass_Preferred
A fluorophore; has been conjugated to streptavidin or secondary antibodies for flow cytometry and used to label bacteria for multi-color FISH; has been coupled to peptides to improve electrospray ionization and MS sensitivity in MALDI-TOF MS; displays absorption/emission maxima of 358/455 nm, respectively
Catalog No. 50-291-1155 Supplier Cayman Chemical Supplier No. 3601710 MG
A fluorophore; has been conjugated to streptavidin or secondary antibodies for flow cytometry and used to label bacteria for multi-color FISH; has been coupled to peptides to improve electrospray ionization and MS sensitivity in MALDI-TOF MS; displays absorption/emission maxima of 358/455 nm, respectively