Learn More
Medchemexpress LLC 6,9-Dioxa-3,12-diazatetradecanedioic acid, 3,12-bis[2-[(acetyloxy)methoxy]-2-oxoethyl]-, 1,14-bis[(acetyloxy)methyl] ester | 99590-86-0 | CB0129710 | 95.0% | 25 MG
SDP
Catalog No. 5000412650
Encompass_Preferred
EGTA-AM is a cell-permeable calcium chelator, typically appearing as a colorless to light yellow liquid. It is supplied with a purity of 95.0% and is designed for high-speed applications in laboratory settings.
  • Consistent with structure confirmed by 1H NMR spectrum
  • Consistent with structure confirmed by MS
  • Store at -20°C, protect from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light
Catalog No. 50-004-12650 Supplier Medchemexpress LLC Supplier No. 25MGHYD0973
EGTA-AM is a cell-permeable calcium chelator, typically appearing as a colorless to light yellow liquid. It is supplied with a purity of 95.0% and is designed for high-speed applications in laboratory settings.
  • Consistent with structure confirmed by 1H NMR spectrum
  • Consistent with structure confirmed by MS
  • Store at -20°C, protect from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light