Learn More
Medchemexpress LLC 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis[2-[(acetyloxy)methoxy]-2-oxoethyl]- | 99590-86-0 | MFCD00077339 | 95.0% | 668.60 | C26H40N2O18 | 10 MG
SDP
Catalog No. 5000297672
Encompass_Preferred
EGTA-AM is the acetoxymethyl ester of EGTA, a membrane-permeable calcium chelator used to deliver EGTA into cells to buffer intracellular Ca2+. It is commonly applied in cell-based assays to reduce cytosolic calcium with relatively slow chelation kinetics compared with fast chelators.
  • Membrane-permeable acetoxymethyl ester for passive cell loading.
  • Buffers intracellular Ca2+ with slow chelating dynamics.
  • Suitable for cell-based assays requiring gradual calcium reduction.
  • Provided as a solid for stock solution preparation.
  • Molecular formula C26H40N2O18; molecular weight 668.60.
Catalog No. 50-002-97672 Supplier Medchemexpress LLC Supplier No. HYD097310MG
EGTA-AM is the acetoxymethyl ester of EGTA, a membrane-permeable calcium chelator used to deliver EGTA into cells to buffer intracellular Ca2+. It is commonly applied in cell-based assays to reduce cytosolic calcium with relatively slow chelation kinetics compared with fast chelators.
  • Membrane-permeable acetoxymethyl ester for passive cell loading.
  • Buffers intracellular Ca2+ with slow chelating dynamics.
  • Suitable for cell-based assays requiring gradual calcium reduction.
  • Provided as a solid for stock solution preparation.
  • Molecular formula C26H40N2O18; molecular weight 668.60.