Learn More
AdipoGen 6-Amino-8-trifluoromethylphenanthridine (5 mg)
SDP
Supplier:  AdipoGen AGMRC0031M005
Encompass_Preferred
Chemical. CAS: 651055-83-3. Formula: C14H9F3N2. MW: 262.2. Potent antiprion agent. Higher activity than 6-Aminophenanthridine (Prod. No. MC-C0029). Ribosome-borne protein folding activity (RPFA) inhibitor. Binds to the ribosomal RNA and inhibits specifically the protein folding activity of the ribosome. Inhibitor of protein aggregation.
Catalog No. 50-205-4252